Learn More
Biomatik Corporation Human Low Density Lipoprotein Receptor Related Protein 1 (LRP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05680
Encompass_Preferred
The Human Low Density Lipoprotein Receptor Related Protein 1 (LRP1) ELISA Kit allows for quantitative determination of Low Density Lipoprotein Receptor Related Protein 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 28.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q07954. Cross Activity: No significant cross-reactivity or interference between Low Density Lipoprotein Receptor Related Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3152