Learn More
Biomatik Corporation Human OxHDL (Oxidized high-density lipoprotein) ELISA Kit
Supplier:  Biomatik Corporation EKF59487
Encompass_Preferred
Human OxHDL (Oxidized high-density lipoprotein) ELISA Kit for detecting OxHDL in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02647; P02652. Target Synonyms: Oxidized high-density lipoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8268