Learn More
Biomatik Corporation Mouse Luteinizing Hormone (LH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05693
Encompass_Preferred
Mouse Luteinizing Hormone (LH) ELISA Kit for detecting LH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 370.4-30,000pg/ml. Sensitivity: 145.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01216; O09108. Gene ID: 12640. Target Synonyms: ICSH; Lutropin; Interstitial Cell Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3155