Learn More
Biomatik Corporation Hamster IL-2 (Interleukin 2) ELISA Kit
Supplier:  Biomatik Corporation EKF59541
Encompass_Preferred
Hamster IL-2 (Interleukin 2) ELISA Kit for detecting IL-2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Hamster. Detection Range: 12.5-800pg/ml. Sensitivity: 7.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: G3HTL8. Target Synonyms: IL-2; Interleukin 2; IL2; TCGF; Lymphokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8271