Learn More
Biomatik Corporation Rat Luteinizing Hormone (LH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05694
Encompass_Preferred
Rat Luteinizing Hormone (LH) ELISA Kit for detecting LH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 98.77-8,000pg/ml. Sensitivity: 37.59pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P16235. Gene ID: 25477. Target Synonyms: ICSH; Lutropin; Interstitial Cell Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3156