Learn More
Biomatik Corporation Mouse Lymphocyte Antigen 75 (LY75) ELISA Kit, 96 tests
Catalog No. 501503158
Encompass_Preferred
Mouse Lymphocyte Antigen 75 (LY75) ELISA Kit for detecting LY75 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60767. Gene ID: 17076. Target Synonyms: CD205; LY75; CLEC13B; DEC-205; GP200-MR6; C-type lectin domain family 13 member B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3158 Supplier Biomatik Corporation Supplier No. EKU05698
Mouse Lymphocyte Antigen 75 (LY75) ELISA Kit for detecting LY75 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60767. Gene ID: 17076. Target Synonyms: CD205; LY75; CLEC13B; DEC-205; GP200-MR6; C-type lectin domain family 13 member B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.