Learn More
Biomatik Corporation Human Lymphocyte Antigen 96 (LY96) ELISA Kit, 96 tests
Catalog No. 501503159
Encompass_Preferred
The Human Lymphocyte Antigen 96 (LY96) ELISA Kit allows for quantitative determination of Lymphocyte Antigen 96 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y6Y9. Cross Activity: No significant cross-reactivity or interference between Lymphocyte Antigen 96 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3159 Supplier Biomatik Corporation Supplier No. EKU05700
The Human Lymphocyte Antigen 96 (LY96) ELISA Kit allows for quantitative determination of Lymphocyte Antigen 96 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y6Y9. Cross Activity: No significant cross-reactivity or interference between Lymphocyte Antigen 96 and analogues was observed. Restriction: For Research Use Only.