Learn More
Biomatik Corporation Human Lymphocyte Function Associated Antigen 3 (LFA3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05705
Encompass_Preferred
The Human Lymphocyte Function Associated Antigen 3 (LFA3) ELISA Kit allows for quantitative determination of Lymphocyte Function Associated Antigen 3 in Serum, Plasma, Cell lysatesand other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.50-800 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P19256. Cross Activity: No significant cross-reactivity or interference between Lymphocyte Function Associated Antigen 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3161