Learn More
Biomatik Corporation Mouse Lysophosphatidylcholine Acyltransferase 1 (LPCAT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05715
Encompass_Preferred
Mouse Lysophosphatidylcholine Acyltransferase 1 (LPCAT1) ELISA Kit for detecting LPCAT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3TFD2. Gene ID: 210992. Target Synonyms: AYTL2; PFAAP3; Acyltransferase Like 2; 1-alkylglycerophosphocholine O-acetyltransferase; Phosphonoformate immuno-associated protein 3; Acetyl-CoA:lyso-PAF acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3163