Learn More
Biomatik Corporation Human Humanin (MT-RNR2) ELISA Kit
Supplier:  Biomatik Corporation EKF60022
Encompass_Preferred
Human Humanin (MT-RNR2) ELISA Kit for detecting MT-RNR2; HN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.146-600pg/ml. Sensitivity: 0.12pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: Q8IVG9. Target Synonyms: MT-RNR2; HN; Humanin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8292