Learn More
Biomatik Corporation Human Lysophosphatidylcholine Acyltransferase 3 (LPCAT3) ELISA Kit, 96 tests
Catalog No. 501503164
Encompass_Preferred
The Human Lysophosphatidylcholine Acyltransferase 3 (LPCAT3) ELISA Kit allows for quantitative determination of Lysophosphatidylcholine Acyltransferase 3 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.221 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6P1A2. Cross Activity: No significant cross-reactivity or interference between Lysophosphatidylcholine Acyltransferase 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3164 Supplier Biomatik Corporation Supplier No. EKU05718
The Human Lysophosphatidylcholine Acyltransferase 3 (LPCAT3) ELISA Kit allows for quantitative determination of Lysophosphatidylcholine Acyltransferase 3 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.221 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6P1A2. Cross Activity: No significant cross-reactivity or interference between Lysophosphatidylcholine Acyltransferase 3 and analogues was observed. Restriction: For Research Use Only.