Learn More
Biomatik Corporation Mouse Lysophosphatidylcholine Acyltransferase 3 (LPCAT3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05719
Encompass_Preferred
Mouse Lysophosphatidylcholine Acyltransferase 3 (LPCAT3) ELISA Kit for detecting LPCAT3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91V01. Gene ID: 14792. Target Synonyms: MBOAT5; C3F; OACT5; Nessy; LPSAT; Membrane Bound O-AcylTransferase Domain Containing 5; Lysophospholipid Acyltransferase 5; Lysophosphatidylserine acyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3165