Learn More
Biomatik Corporation Human Lysosomal Associated Membrane Protein 2 (LAMP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05723
Encompass_Preferred
The Human Lysosomal Associated Membrane Protein 2 (LAMP2) ELISA Kit allows for quantitative determination of Lysosomal Associated Membrane Protein 2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4,000 pg/mL. Sensitivity: 24.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13473. Cross Activity: No significant cross-reactivity or interference between Lysosomal Associated Membrane Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3167