Learn More
Biomatik Corporation Cattle Lysozyme (LZM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05724
Encompass_Preferred
The Cattle Lysozyme (LZM) ELISA Kit allows for quantitative determination of Lysozyme in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.131 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04421. Cross Activity: No significant cross-reactivity or interference between Lysozyme and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3168