Learn More
Biomatik Corporation Gallus Lysozyme (LZM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05725
Encompass_Preferred
The Gallus Lysozyme (LZM) ELISA Kit allows for quantitative determination of Lysozyme in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.262 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00698. Cross Activity: No significant cross-reactivity or interference between Lysozyme and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3169