Learn More
Biomatik Corporation Human Lysyl Oxidase (LOX) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05728
Encompass_Preferred
The Human Lysyl Oxidase (LOX) ELISA Kit allows for quantitative determination of Lysyl Oxidase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P28300. Cross Activity: No significant cross-reactivity or interference between Lysyl Oxidase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3172