Learn More
Biomatik Corporation Human 5-NT (5-Nucleotidase) ELISA Kit
Supplier:  Biomatik Corporation EKF60398
Encompass_Preferred
Human 5-NT (5-Nucleotidase) ELISA Kit for detecting 5-NT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21589. Target Synonyms: 5-NT; NT5E; 5'-NT; CD73; E5NT; eNT; NTE; eN; NT; NT5; 5' nucleotidase; CD73; CD73 antigen; Ecto-5'-nucleotidase; 5'-nucleotidase; Purine 5-Prime-Nucleotidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8332