Learn More
Biomatik Corporation Mouse Lysyl Oxidase Like Protein 2 (LOXL2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05733
Encompass_Preferred
Mouse Lysyl Oxidase Like Protein 2 (LOXL2) ELISA Kit for detecting LOXL2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58022. Gene ID: 94352. Target Synonyms: LOR2; WS9-14; Lysyl oxidase homolog 2; Lysyl oxidase-related protein 2; Lysyl oxidase-related protein WS9-14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3176