Learn More
Biomatik Corporation Rat Lysyl Oxidase Like Protein 2 (LOXL2) ELISA Kit, 96 tests
Catalog No. 501503177
Encompass_Preferred
Rat Lysyl Oxidase Like Protein 2 (LOXL2) ELISA Kit for detecting LOXL2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B5DF27. Gene ID: 290350. Target Synonyms: LOR2; WS9-14; Lysyl oxidase homolog 2; Lysyl oxidase-related protein 2; Lysyl oxidase-related protein WS9-14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3177 Supplier Biomatik Corporation Supplier No. EKU05734
Rat Lysyl Oxidase Like Protein 2 (LOXL2) ELISA Kit for detecting LOXL2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B5DF27. Gene ID: 290350. Target Synonyms: LOR2; WS9-14; Lysyl oxidase homolog 2; Lysyl oxidase-related protein 2; Lysyl oxidase-related protein WS9-14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.