Learn More
Biomatik Corporation Mouse IL-5 (Interleukin 5) ELISA Kit
Supplier:  Biomatik Corporation EKF60408
Encompass_Preferred
Mouse IL-5 (Interleukin 5) ELISA Kit for detecting IL-5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04401. Target Synonyms: IL-5; IL5; EDF; TRF; E-CSF; ESGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8340