Learn More
Biomatik Corporation Mouse Lysyl tRNA Synthetase (KARS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05738
Encompass_Preferred
Mouse Lysyl tRNA Synthetase (KARS) ELISA Kit for detecting KARS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99MN1. Gene ID: 85305. Target Synonyms: KARS2; LysRS; Lysine tRNA Ligase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3178