Learn More
Biomatik Corporation Mouse Macrophage Inflammatory Protein 1 Alpha (MIP1a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05745
Encompass_Preferred
Mouse Macrophage Inflammatory Protein 1 Alpha (MIP1a) ELISA Kit for detecting MIP1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10855. Gene ID: 20302. Target Synonyms: CCL3; MIP1-A; SCYA3; Chemokine C-C-Motif Ligand 3; Small Inducible Cytokine A3; Homologous To Mouse Mip-1a; Tonsillar Lymphocyte LD78 Alpha Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3181