Learn More
Biomatik Corporation Human Macrophage Inflammatory Protein 3 Alpha (MIP3a) ELISA Kit, 96 tests
Catalog No. 501503185
Encompass_Preferred
The Human Macrophage Inflammatory Protein 3 Alpha (MIP3a) ELISA Kit allows for quantitative determination of Macrophage Inflammatory Protein 3 Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P78556. Cross Activity: No significant cross-reactivity or interference between Macrophage Inflammatory Protein 3 Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3185 Supplier Biomatik Corporation Supplier No. EKU05752
The Human Macrophage Inflammatory Protein 3 Alpha (MIP3a) ELISA Kit allows for quantitative determination of Macrophage Inflammatory Protein 3 Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P78556. Cross Activity: No significant cross-reactivity or interference between Macrophage Inflammatory Protein 3 Alpha and analogues was observed. Restriction: For Research Use Only.