Learn More
Biomatik Corporation Human CD52 (CAMPATH-1 antigen) ELISA Kit
Supplier:  Biomatik Corporation EKF60462
Encompass_Preferred
Human CD52 (CAMPATH-1 antigen) ELISA Kit for detecting CD52 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P31358. Target Synonyms: CD52; CAMPATH-1 antigen; CDW52; HE5; Human epididymis-specific protein 5; Epididymal secretory protein E5; Cambridge pathology 1 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8362