Learn More
Biomatik Corporation Human Macrophage Migration Inhibitory Factor (MIF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05761
Encompass_Preferred
The Human Macrophage Migration Inhibitory Factor (MIF) ELISA Kit allows for quantitative determination of Macrophage Migration Inhibitory Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.128 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P14174. Cross Activity: No significant cross-reactivity or interference between Macrophage Migration Inhibitory Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3188