Learn More
Biomatik Corporation Rat Macrophage Migration Inhibitory Factor (MIF) ELISA Kit, 96 tests
Catalog No. 501503190
Encompass_Preferred
Rat Macrophage Migration Inhibitory Factor (MIF) ELISA Kit for detecting MIF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30904. Gene ID: 81683. Target Synonyms: GIF; GLIF; MMIF; Glycosylation-Inhibiting Factor; L-dopachrome isomerase; L-dopachrome tautomerase; Phenylpyruvate tautomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3190 Supplier Biomatik Corporation Supplier No. EKU05763
Rat Macrophage Migration Inhibitory Factor (MIF) ELISA Kit for detecting MIF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30904. Gene ID: 81683. Target Synonyms: GIF; GLIF; MMIF; Glycosylation-Inhibiting Factor; L-dopachrome isomerase; L-dopachrome tautomerase; Phenylpyruvate tautomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.