Learn More
Biomatik Corporation Mouse Major Basic Protein (MBP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05769
Encompass_Preferred
Mouse Major Basic Protein (MBP) ELISA Kit for detecting MBP in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61878. Gene ID: 19074. Target Synonyms: PRG2; BMPG; Proteoglycan 2; Bone Marrow,Natural Killer Cell Activator; Eosinophil Granule Major Basic Protein; Proteoglycan 2,Bone Marrow; Pregnancy-associated major basic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3193