Learn More
Biomatik Corporation Human Smad3/MADH3 (Mothers Against Decapentaplegic Homolog 3) ELISA Kit
Supplier:  Biomatik Corporation EKF60672
Encompass_Preferred
Human Smad3/MADH3 (Mothers Against Decapentaplegic Homolog 3) ELISA Kit for detecting Smad3; MADH3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P84022. Target Synonyms: Smad3; MADH3; JV15-2; SMAD family member 3; hMAD-3; hSMAD3; MAD homolog 3; mad protein homolog; MAD; mothers against decapentaplegic homolog 3; MAD; mothers against decapentaplegic homolog 3; Drosophila; mad3; Mad3; MADH3mad homolog JV15-2; mothers against decapentaplegic homolog 3; Mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD family member 3HSPC193; SMAD; mothers against DPP homolog 3; SMAD; mothers against DPP homolog 3; Drosophila; Smad3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8382