Learn More
Biomatik Corporation Human Major Histocompatibility Complex Class I E (MHCE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05772
Encompass_Preferred
The Human Major Histocompatibility Complex Class I E (MHCE) ELISA Kit allows for quantitative determination of Major Histocompatibility Complex Class I E in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.25-400 pg/mL. Sensitivity: 2.41 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13747. Cross Activity: No significant cross-reactivity or interference between Major Histocompatibility Complex Class I E and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3196