Learn More
Biomatik Corporation Malondialdehyde (MDA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05783
Encompass_Preferred
Malondialdehyde (MDA) ELISA Kit for detecting MDA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 2.47-200ng/ml. Sensitivity: 0.89ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3200