Learn More
Biomatik Corporation Porcine FSH (Follicle-Stimulating Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF60716
Encompass_Preferred
Porcine FSH (Follicle-Stimulating Hormone) ELISA Kit for detecting FSH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 23.438-1500ng/ml. Sensitivity: 14.063ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P01228. Target Synonyms: FSH; Follicle-Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8401