Learn More
Biomatik Corporation Mouse Mannose Associated Serine Protease 1 (MASP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05786
Encompass_Preferred
Mouse Mannose Associated Serine Protease 1 (MASP1) ELISA Kit for detecting MASP1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98064. Gene ID: 17174. Target Synonyms: CRARF; PRSS5; Mannan-Binding Lectin Serine Peptidase 1; Ra-reactive factor serine protease p100; Complement-activating component of Ra-reactive factor; Serine protease 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3202