Learn More
Biomatik Corporation Porcine OC/BGP (Osteocalcin/Bone Gla Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF60720
Encompass_Preferred
Porcine OC/BGP (Osteocalcin/Bone Gla Protein) ELISA Kit for detecting OC; BGP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8HYY9. Target Synonyms: OC; Osteocalcin; BGLAP; BGP; OCN; bone gamma-carboxyglutamate; gla protein; Bone Gla protein; Gamma-carboxyglutamic acid-containing protein; Bone gamma-carboxyglutamic Acid Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8405