Learn More
Biomatik Corporation Gallus Mannose Binding Lectin (MBL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05788
Encompass_Preferred
The Gallus Mannose Binding Lectin (MBL) ELISA Kit allows for quantitative determination of Mannose Binding Lectin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q98TA4. Cross Activity: No significant cross-reactivity or interference between Mannose Binding Lectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3204