Learn More
Biomatik Corporation Human MARCKS Related Protein (MARCKSL1) ELISA Kit, 96 tests
Catalog No. 501503208
Encompass_Preferred
The Human MARCKS Related Protein (MARCKSL1) ELISA Kit allows for quantitative determination of MARCKS Related Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49006. Cross Activity: No significant cross-reactivity or interference between MARCKS Related Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3208 Supplier Biomatik Corporation Supplier No. EKU05803
The Human MARCKS Related Protein (MARCKSL1) ELISA Kit allows for quantitative determination of MARCKS Related Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P49006. Cross Activity: No significant cross-reactivity or interference between MARCKS Related Protein and analogues was observed. Restriction: For Research Use Only.