Learn More
Biomatik Corporation Human Matrix Gla Protein (MGP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05813
Encompass_Preferred
The Human Matrix Gla Protein (MGP) ELISA Kit allows for quantitative determination of Matrix Gla Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.254 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08493. Cross Activity: No significant cross-reactivity or interference between Matrix Gla Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3211