Learn More
Biomatik Corporation Mouse Matrix Gla Protein (MGP) ELISA Kit, 96 tests
Catalog No. 501503212
Encompass_Preferred
Mouse Matrix Gla Protein (MGP) ELISA Kit for detecting MGP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19788. Gene ID: 17313. Target Synonyms: MGLAP; GIG36; Cell growth-inhibiting gene 36 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3212 Supplier Biomatik Corporation Supplier No. EKU05814
Mouse Matrix Gla Protein (MGP) ELISA Kit for detecting MGP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.47-30ng/ml. Sensitivity: 0.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19788. Gene ID: 17313. Target Synonyms: MGLAP; GIG36; Cell growth-inhibiting gene 36 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.