Learn More
Biomatik Corporation Human MCFD2 (Multiple coagulation factor deficiency protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF60766
Encompass_Preferred
Human MCFD2 (Multiple coagulation factor deficiency protein 2) ELISA Kit for detecting MCFD2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8NI22. Target Synonyms: MCFD2; Multiple coagulation factor deficiency protein 2; Neural stem cell-derived neuronal survival protein; SDNSF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8435