Learn More
Biomatik Corporation Human FCER1A (High affinity immunoglobulin epsilon receptor subunit alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60771
Encompass_Preferred
Human FCER1A (High affinity immunoglobulin epsilon receptor subunit alpha) ELISA Kit for detecting FCER1A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12319. Target Synonyms: FCER1A; FCE1A; FcERI; Fc-epsilon RI-alpha; IgE Fc receptor subunit alpha;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8439