Learn More
Biomatik Corporation Human Matrix Metalloproteinase 10 (MMP10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05824
Encompass_Preferred
The Human Matrix Metalloproteinase 10 (MMP10) ELISA Kit allows for quantitative determination of Matrix Metalloproteinase 10 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78.12-5000 pg/mL. Sensitivity: 34 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09238. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 10 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3216