Learn More
Biomatik Corporation Human Matrix Metalloproteinase 11 (MMP11) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05828
Encompass_Preferred
The Human Matrix Metalloproteinase 11 (MMP11) ELISA Kit allows for quantitative determination of Matrix Metalloproteinase 11 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.27 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24347. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 11 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3217