Learn More
Biomatik Corporation Mouse OVA sIgG (Ovalbumin Specific IgG) ELISA Kit
Supplier:  Biomatik Corporation EKF60781
Encompass_Preferred
Mouse OVA sIgG (Ovalbumin Specific IgG) ELISA Kit for detecting OVA sIgG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Indirect ELISA. Assay Time: 3h, 30 min. Accession Number: P01012. Target Synonyms: OVA sIgG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8446