Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 12 (MMP12) ELISA Kit, 96 tests
Catalog No. 501503218
Encompass_Preferred
Mouse Matrix Metalloproteinase 12 (MMP12) ELISA Kit for detecting MMP12 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34960. Gene ID: 17381. Target Synonyms: MME; HME; ME; Macrophage Metalloelastase; Macrophage Elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3218 Supplier Biomatik Corporation Supplier No. EKU05830
Mouse Matrix Metalloproteinase 12 (MMP12) ELISA Kit for detecting MMP12 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34960. Gene ID: 17381. Target Synonyms: MME; HME; ME; Macrophage Metalloelastase; Macrophage Elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.