Learn More
Biomatik Corporation Monkey IFN-γ (Interferon Gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF60785
Encompass_Preferred
Monkey IFN-γ (Interferon Gamma) ELISA Kit for detecting IFN-γ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P63309. Target Synonyms: IFN-γ; IFNG; IFG; IFI; Type II Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8450