Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 12 (MMP12) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05831
Encompass_Preferred
Rat Matrix Metalloproteinase 12 (MMP12) ELISA Kit for detecting MMP12 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63341. Gene ID: 117033. Target Synonyms: MME; HME; ME; Macrophage Metalloelastase; Macrophage Elastase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3219