Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 13 (MMP13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05837
Encompass_Preferred
Mouse Matrix Metalloproteinase 13 (MMP13) ELISA Kit for detecting MMP13 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33435. Gene ID: 17386. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3221