Learn More
Biomatik Corporation Pig Matrix Metalloproteinase 13 (MMP13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05838
Encompass_Preferred
Pig Matrix Metalloproteinase 13 (MMP13) ELISA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1SV56. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3222