Learn More
Biomatik Corporation Rat Matrix Metalloproteinase 13 (MMP13) ELISA Kit, 96 tests
Catalog No. 501503223
Encompass_Preferred
Rat Matrix Metalloproteinase 13 (MMP13) ELISA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23097. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3223 Supplier Biomatik Corporation Supplier No. EKU05839
Rat Matrix Metalloproteinase 13 (MMP13) ELISA Kit for detecting MMP13 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23097. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.