Learn More
Biomatik Corporation Rabbit Matrix Metalloproteinase 13 (MMP13) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05840
Encompass_Preferred
Rabbit Matrix Metalloproteinase 13 (MMP13) ELISA Kit for detecting MMP13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O62806. Gene ID: 100008685. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3224