Learn More
Biomatik Corporation Hamster IL-1b (Interleukin-1b) ELISA Kit
Supplier:  Biomatik Corporation EKF60820
Encompass_Preferred
Hamster IL-1b (Interleukin-1b) ELISA Kit for detecting IL-1b in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Hamster. Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9WVQ9. Target Synonyms: IL-1b; Interleukin-1b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8478